Learn More
PHF21B Monoclonal Antibody (OTI5H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503284
Description
PHF21B (PHD finger protein 21B), also known as PHF4, is a 531 amino acid protein that contains one PHD-type zinc finger and is expressed as multiple alternatively spliced isoforms. The gene encoding PHF21B maps to human chromosome 22, which houses over 500 genes and is the second smallest human chromosome. Mutations in several of the genes that map to chromosome 22 are involved in the development of Phelan-McDermid syndrome, neurofibromatosis type 2, autism and schizophrenia.Specifications
| PHF21B | |
| Monoclonal | |
| 0.41 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96EK2 | |
| PHF21B | |
| Full length human recombit protein of human PHF21B produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5H9 | |
| Unconjugated | |
| PHF21B | |
| BHC80L, PHF4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 112885 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.