missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00€ - 572.00€
Specifications
| Antigen | PHIP |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18407521
|
Novus Biologicals
NBP2-33886-25ul |
25 μL |
280.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18152105
|
Novus Biologicals
NBP2-33886 |
0.1 mL |
572.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PHIP Polyclonal specifically detects PHIP in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| PHIP | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q8WWQ0 | |
| 55023 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TEQPSTSSAAKTFITKANASAIPGKTILENSVKHSKALNTLSSPGQSSFSHGTRNNSAKENMEKEKPVKRKMKSSVLPKASTLSKSSA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DCAF14, DDB1 and CUL4 associated factor 14, FLJ20705, FLJ45918, IRS-1 PH domain-binding protein, MGC90216, ndrp, PH-interacting protein, pleckstrin homology domain interacting protein, WD repeat-containing protein 11, WDR11 | |
| PHIP | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title