missing translation for 'onlineSavingsMsg'
Learn More

Phorate Oxon, TRC

Product Code. 30134828
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134828 100 mg 100mg
18080430 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134828 Supplier Toronto Research Chemicals Supplier No. TRCP353510100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2600-69-3
Molecular Formula C7 H17 O3 P S2
Molecular Weight (g/mol) 244.31
Synonym Phosphorothioic acid, O,O-diethyl S-[(ethylthio)methyl] ester, Methanethiol, (ethylthio)-, S-ester with O,O-diethyl phosphorothioate (8CI), O,O-Diethyl S-(ethylthiomethyl) phosphorothioate, Phorate oxon, Phorate oxygen analog, Phorate thiolate analog, Phoratoxon, Thimet oxon, Thimet oxygen analog, Phorate-oxon
IUPAC Name 1-[ethoxy(ethylsulfanylmethylsulfanyl)phosphoryl]oxyethane
SMILES CCOP(=O)(OCC)SCSCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H17O3PS2/c1-4-9-11(8,10-5-2)13-7-12-6-3/h4-7H2,1-3H3
Formula Weight 244.0357
Chemical Name or Material Phorate Oxon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.