missing translation for 'onlineSavingsMsg'
Learn More

Phosalone, TRC

Product Code. 30133536
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133536 10 g 10g
30040375 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133536 Supplier Toronto Research Chemicals Supplier No. TRCP35360010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2310-17-0
Molecular Formula C12 H15 Cl N O4 P S2
Molecular Weight (g/mol) 367.81
Synonym Phosphorodithioic acid, O,O-diethyl ester, S-ester with 6-chloro-3-(mercaptomethyl)-2-benzoxazolinone (7CI,8CI), 3-(O,O-Diethyldithiophosphorylmethyl)-6-chlorobenzoxazolinone, 6-Chloro-3-(O,O-diethyldithiophosphorylmethyl)benzoxazolone, Agria 1060, Benzophosphate, Chipman 11974, Fosalon, Fozolone, KB insectes, NIA 9241, O,O-Diethyl S-[6-chloro-3-(mercaptomethyl)-2-benzoxazolinone]phosphorodithioate, Phosalon, Phosalone, Phosphorodithioic acid O,O-diethyl S-[(6-chloro-2-oxobenzoxazolin-3-yl)methyl] ester, RP 11974, Rubitox, S-(6-Chloro-2-oxobenzoxazolin-3-yl)methyl diethyl phosphorothiolothionate, S-[6-Chloro-3-(mercaptomethyl)-2-benzoxazolinone] O,O-diethyl phosphorodithioate, Zolone, Zolone 35EC, Zolone Flo, Zolone PM, Phosphorodithioic acid, S-[(6-chloro-2-oxo-3(2H)-benzoxazolyl)methyl] O,O-diethyl ester
IUPAC Name 6-chloro-3-(diethoxyphosphinothioylsulfanylmethyl)-1,3-benzoxazol-2-one
SMILES CCOP(=S)(OCC)SCN1C(=O)Oc2cc(Cl)ccc12

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H15ClNO4PS2/c1-3-16-19(20,17-4-2)21-8-14-10-6-5-9(13)7-11(10)18-12(14)15/h5-7H,3-4,8H2,1-2H3
Formula Weight 366.9869
Chemical Name or Material Phosalone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.