Learn More
Phospho-AKT (Thr308) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
135.00€ - 360.00€
Specifications
| Antigen | Phospho-AKT (Thr308) |
|---|---|
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17422373
|
Proteintech
29163-1-AP-20UL |
20 μL |
135.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17412373
|
Proteintech
29163-1-AP-100UL |
100 μL |
360.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-AKT (Thr308) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-AKT (Thr308). This antibody recognizes human antigen. The Phospho-AKT (Thr308) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Phospho-AKT (Thr308) | |
| Polyclonal | |
| Rabbit | |
| 207 | |
| Primary |
| Western Blot, ELISA | |
| Unconjugated | |
| Human | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.