missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-AKT1 (Ser124) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538251
Additional Details : Weight : 0.00900kg
Description
AKT, also known as protein kinase B (PKB), is a 57 kDa serine/threonine protein kinase. There are three mammalian isoforms of Akt: AKT1 (PKB alpha), AKT2 (PKB beta) and AKT3 (PKB gamma) with AKT2 and AKT3 being approximately 82% identical with the AKT1 isoform. Each isoform has a pleckstrin homology (PH) domain, a kinase domain and a carboxy terminal regulatory domain. AKT was originally cloned from the retrovirus AKT8, and is a key regulator of many signal transduction pathways. Its tight control over cell proliferation and cell viability are manifold; overexpression or inappropriate activation of AKT has been seen in many types of cancer.Specifications
Phospho-AKT1 (Ser124) | |
Polyclonal | |
Unconjugated | |
AKT1 | |
RAC-alpha serine/threonine-protein kinase, Protein kinase B, PKB, Protein kinase B alpha, PKB alpha, Proto-oncogene c-Akt, RAC-PK-alpha, AKT1, PKB, RAC, PKB alpha, RAC-beta serine/threonine-protein kinase, Protein kinase Akt-2, Protein kinase B beta, PKB beta, RAC-PK-beta, AKT2, PKB beta, RAC-gamma serine/threonine-protein kinase, Protein kinase Akt-3, Protein kinase B gamma, PKB gamma, RAC-PK-gamma, STK-2, AKT3, PKBG, PKB gamma | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11651, 207, 24185 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P31749, P31750, P47196 | |
AKT1 | |
A synthetic phosphopeptide derived from human Akt around the phosphorylation site of Ser124 (S-G-SP-P-S) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |