missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-c-Cbl (Tyr774) Recombinant Rabbit Monoclonal Antibody (CblY774-R3B8), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
331.00€ - 662.00€
Specifications
| Antigen | Phospho-c-Cbl (Tyr774) |
|---|---|
| Clone | CblY774-R3B8 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17136938
|
Invitrogen™
MA537347 |
20 μL |
331.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17176688
![]() |
Invitrogen™
MA537101 |
200 μL |
662.00€
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cbl proteins are a family of ubiquitin protein ligases (E3s) that negatively regulate signaling by targeting activated tyrosine kinases for degradation. Cbl- c is the most recently cloned member of the Cbl proteins and is expressed only in epithelial cells (the other Cbl proteins are ubiquitously expressed). Cbl-c, like the other mammalian Cbl proteins, can ubiquitinate the activated EGFR and target it for degradation. Through interactions with proteins containing SRC homology-2 (SH2) and SH3 domains, CBL proteins modulate downstream cell signaling.Specifications
| Phospho-c-Cbl (Tyr774) | |
| 0.5 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.10% BSA, 50% glycerol and 0.02% sodium azide | |
| 4732447J05Rik; Cas-Br-M (murine) ecotropic retroviral transforming sequence; Casitas B-lineage lymphoma; casitas B-lineage lymphoma proto-oncogene; CBL; Cbl proto-oncogene; Cbl proto-oncogene, E3 ubiquitin protein ligase; CBL2; Cbl-2; CBL-3; CBLC; c-Cbl; E3 ubiquitin-protein ligase CBL; FRA11B; fragile site, folic acid type, rare, fra(11)(q23.3); NSLL; oncogene CBL2; proto-oncogene c-Cbl; RGD1561386; RING finger protein 55; RING-type E3 ubiquitin transferase CBL; RNF55; RNF57; signal transduction protein CBL | |
| Cbl | |
| IgG κ | |
| Protein A/G | |
| Antibody |
| CblY774-R3B8 | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P22681, P22682 | |
| 12402, 867 | |
| A synthetic phospho-peptide corresponding to residues surrounding Tyr774 of human phospho c-Cbl. | |
| Primary | |
| -20°C | |
| Cbl |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title