missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Cullin 4A (Ser40) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105313
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of CUL4A only when phosphorylated at Ser40.
CUL4A (Cullin 4A) is the ubiquitin ligase component of a multimeric complex involved in the degradation of DNA damage-response proteins.
Specifications
| Phospho-Cullin 4A (Ser40) | |
| Polyclonal | |
| Unconjugated | |
| CUL4A | |
| 2810470J21Rik; AW495282; CUL 4A; Cul4a; CUL-4A; Cul4a protein; cullin 4A; cullin-4A; MGC36573; MGC64071; OTTHUMP00000018753; RGD1563853 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 361181, 8451, 99375 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13619, Q3TCH7 | |
| CUL4A | |
| A synthesized peptide derived from human CUL4A(Accession Q13619), corresponding to amino acid residues around phosphorylated Ser40. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction