missing translation for 'onlineSavingsMsg'
Learn More

Phospho-DNM1L (Ser616) Recombinant Rabbit Monoclonal Antibody (PSH06-77)
GREENER_CHOICE

Phospho-DNM1L (Ser616) Recombinant Rabbit Monoclonal Antibody (PSH06-77)

Brand:  Invitrogen MA556582

Product Code. 30275221

  • 446.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Phospho-DNM1L (Ser616) Recombinant Monoclonal Antibody for WB, ICC/IF, IHC (P), Flow

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-DNM1L (Ser616)
Recombinant Monoclonal
1 mg/mL
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
O00429, O35303, Q8K1M6
DNM1L
Synthetic phospho-peptide corresponding to residues surrounding Ser616 of human DRP1.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
PSH06-77
Unconjugated
DNM1L
6330417M19Rik; AI450666; C-terminal region; Dlp1; dnm1l; Dnm1p/Vps1p-like protein; Dnml1; Dnmlp1; Drp1; DVLP; DYMPLE; dynamin 1 like; dynamin 1-like; dynamin family member proline-rich carboxyl-terminal domain less; Dynamin-1-like protein; Dynamin-1-like protein-like protein; Dynamin-like protein; dynamin-like protein 1; dynamin-like protein 4; Dynamin-like protein IV; dynamin-related protein 1; DYNIV 11; DYNIV-11; EC 3.6.5.5; EGK_03514; EMPF; FLJ41912; HDYNIV; N-terminal region; python; similar to dynamin 1-like; VPS1; zgc:66163
Rabbit
Protein A
RUO
10059, 114114, 74006
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.