Learn More
Phospho(enol)pyruvic acid cyclohexylammonium salt, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 10526-80-4 |
| Molecular Formula | C9H18NO6P |
| MDL Number | MFCD00036375 |
| Quantity | 1 g |
| SMILES | NC1CCCCC1.C=C(OP(O)(O)=O)C(O)=O |
| Molecular Weight (g/mol) | 267.22 |
| Percent Purity | 98% |
| Chemical Name or Material | Phospho(enol)pyruvic acid cyclohexylammonium salt |