missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-EphB6 (Tyr651) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5106137
Additional Details : Weight : 0.01000kg
Description
Antibody detects endogenous levels of EPHB6 only when phosphorylated at Tyr651.
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The ephrin receptor encoded by this gene lacks the kinase activity of most receptor tyrosine kinases and binds to ephrin-B ligands.Specifications
Phospho-EphB6 (Tyr651) | |
Polyclonal | |
Unconjugated | |
EPHB6 | |
Cekl; chicken eph/elk receptor-like protein; EPB6; EPH receptor B6; EPHB6; EphB6v; Epherin B6; Epherin receptor B6; ephrin type A receptor; ephrin type-B receptor 6; HEP; human kinase-defective Eph-family receptor protein; MEP; MGC129910; MGC129911; Tyrosine-protein kinase-defective receptor EPH-6 | |
Rabbit | |
sequential chromatography | |
RUO | |
13848, 2051, 312275 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O08644, O15197, P0C0K7 | |
EPHB6 | |
A synthesized peptide derived from human EPHB6(Accession O15197), corresponding to amino acid residues around phosphorylated Tyr651. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |