missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-GSK3 alpha/beta (Ser21, Ser9) Recombinant Rabbit Monoclonal Antibody (2J1P4)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551516
This item is not returnable.
View return policy
Description
Immunogen sequence: TSSFA.
Glycogen synthase kinase-3 (GSK3) is a protein kinase that was originally identified as a regulator of glycogen synthase, a key enzyme in glycogen metabolism. Since then, it has been shown to be involved in the regulation of a diverse array of cellular functions, including protein synthesis, cell proliferation, cell differentiation, microtubule assembly/disassembly, and apoptosis. GSK3's substrate specificity is unique in that phosphorylation of the substrate only occurs if a phosphoserine or phosphotyrosine is present - four residues C-terminal to the site of GSK phosphorylation. There exists two isoforms of GSK3, GSK3 alpha and GSK3 beta, and they are strictly regulated via phosphorylation. Phosphorylation of GSK3 beta on Ser9 (Ser21 in GSK3alpha) by protein kinase B (PKB) causes its inactivation and is the primary mechanism responsible for growth factor inhibition of this kinase. Activation of GSK3 beta is dependent upon the phosphorylation of Tyr216 (Tyr279 in GSK3 alpha). Upon activation, it has been shown to phosphorylate a number of different cellular proteins, including p53, c-Myc, c-Jun, heat shock factor-1 (HSF-1), and cyclin D1. GSK3 beta also has been shown to phosphorylate aberrant sites on the microtubule associated protein tau, which is critical for the progression of Alzheimer's disease.
Specifications
| Phospho-GSK3 alpha/beta (Ser21, Ser9) | |
| Recombinant Monoclonal | |
| 0.83 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q2NL51, Q9WV60 | |
| GSK3A, GSK3B | |
| A synthetic phosphorylated peptide around S21&S9 of human GSK3 alpha/GSK3 beta (NP_063937.2/NP_002084.2). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| Liquid |
| ELISA, Western Blot | |
| 2J1P4 | |
| Unconjugated | |
| GSK3A | |
| 2700086H06Rik; 7330414F15Rik; 8430431H08Rik; C86142; FA; factor A; glycogen synthase kinase 3 alpha; glycogen synthase kinase 3 beta; glycogen synthase kinase-3 alpha; Glycogen synthase kinase-3 beta; GSK3; GSK-3; GSK-3 alpha; GSK-3 beta; Gsk3a; gsk-3-alpha; Gsk3b; GSK-3beta; GSK3beta isoform; serine/threonine-protein kinase GSK3A; Serine/threonine-protein kinase GSK3B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56637, 606496 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction