missing translation for 'onlineSavingsMsg'
Learn More

Phospho-IRF3 (Ser396) Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 29528-1-AP-20UL

 View more versions of this product

Product Code. 30236919

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

This item is not returnable. View return policy

Description

Description

The Phospho-IRF3 (Ser396) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-IRF3 (Ser396). This antibody recognizes human antigen. The Phospho-IRF3 (Ser396) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications

Specifications

IRF3 (Ser396)
400 μg/mL
Rabbit
Antigen affinity purification
Primary
Human
50-60 kDa
Liquid
Western Blot, ELISA
Unconjugated
Peptide
20 μL
3661
Store at -20°C. Stable for one year after shipment.
Polyclonal Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.