missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-MCM2 (Ser139) Recombinant Rabbit Monoclonal Antibody (MCM2S139-B12), FITC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537427
This item is not returnable.
View return policy
Description
MCM2 (Mini chromosome maintenance protein 2) is involved in regulating DNA replication. MCM2 (also called CDCL1, mitotin and BM28), is a human nuclear protein that is crucial in the cell cycle, being involved in the onset of DNA replication and cell division. It is similar to members of the family of early S-phase proteins. MCM2 has been mapped to 3q21. From its localization, CDCL1 became a candidate for an oncogene affected by chromosomal breaks in acute myeloid leukemia (AML).
Specifications
| Phospho-MCM2 (Ser139) | |
| Recombinant Monoclonal | |
| FITC | |
| MCM2 | |
| AA959861; AW476101; BM28; CCNL1; cdc19; CDCL1; cell devision cycle-like 1; cyclin-like 1; D3S3194; DFNA70; DNA replication licensing factor MCM2; Kiaa0030; MCM DNA helicase complex subunit MCM2; Mcm2; Mcmd2; MGC10606; mini chromosome maintenance deficient 2; minichromosome maintenance complex component 2; minichromosome maintenance deficient 2 (mitotin); minichromosome maintenance deficient 2 mitotin; minichromosome maintenance deficient 2 mitotin (S. cerevisiae); Minichromosome maintenance protein 2; Minichromosome maintenance protein 2 homolog; MITOTIN; mKIAA0030; Nuclear protein BM28; YBL023C; YBL0438 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 17216, 312538, 4171 | |
| 4°C, store in dark | |
| Liquid |
| Flow Cytometry | |
| MCM2S139-B12 | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| P49736, P97310 | |
| MCM2 | |
| A synthetic phospho-peptide corresponding to residues surrounding Ser139 of human phospho MCM2. | |
| 10 Tests | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction