missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-MKK7 (Ser271, Thr275) Recombinant Rabbit Monoclonal Antibody (MKK7S271T275-R4F9), FITC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
202.00€ - 508.00€
Specifications
| Antigen | Phospho-MKK7 (Ser271, Thr275) |
|---|---|
| Clone | MKK7S271T275-R4F9 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17116878
|
Invitrogen™
MA537285 |
10 Tests |
202.00€
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17136598
![]() |
Invitrogen™
MA537007 |
100 Tests |
508.00€
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a dual specificity protein kinase that belongs to the MAP kinase family. This kinase specifically activates MAPK8/JNK1 and MAPK9/JNK2, and this kinase itself is phosphorylated and activated by MAP kinase kinases including MAP3K1/MEKK1, MAP3K2/MEKK2, MAP3K3/MEKK5, and MAP4K2/GCK. This kinase is involved in the signal transduction mediating the cell responses to proinflammatory cytokines, and environmental stresses. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found, but only one transcript variant has been supported and defined.Specifications
| Phospho-MKK7 (Ser271, Thr275) | |
| Flow Cytometry | |
| FITC | |
| Rabbit | |
| Human | |
| O14733 | |
| 5609 | |
| A synthetic phospho-peptide corresponding to residues surrounding Ser271 and Thr275 of human MKK7. | |
| Primary | |
| 4°C, store in dark | |
| MAP2K7 |
| MKK7S271T275-R4F9 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| 2; 5930412N11Rik; c-Jun N-terminal kinase kinase 2; Dual specificity mitogen-activated protein kinase kinase 7; EMBL:AAX61178.1}; JN; JNK kinase 2; JNK-activating kinase 2; JNKK; JNKK 2; Jnkk2; MAP kinase kinase 7; MAP2K4; map2k7; map2k7 {ECO:0000312; MAPK/ERK kinase 4; MAPK/ERK kinase 7; MAPKK 4; MAPKK 7; MAPKK7; MEK; MEK 4; MEK 7; Mek7; mitogen activated protein kinase kinase 7; mitogen-activated protein kinase kinase 7; Mkk7; PRKMK7; protein kinase, mitogen activated, kinase 7; SAPK kinase 1; SAPK kinase 4; SAPK/ERK kinase 1; SAPKK1; SAPKK-1; SAPKK4; SAPKK-4; SEK1; sek2; SKK4; Stress-activated protein kinase kinase 4 | |
| MAP2K7 | |
| IgG κ | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title