missing translation for 'onlineSavingsMsg'
Learn More

Phospho-P53 (Ser15) Mouse, anti-Human, Unconjugated, Clone: 1H6G1, Proteintech

Mouse Monoclonal Antibody

135.00€ - 360.00€

Specifications

Antigen Phospho-P53 (Ser15)
Clone 1H6G1
Concentration 1000 μg/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17251476
View Documents
Proteintech
67826-1-IG-20UL
20 μL
135.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17241476
View Documents
Proteintech
67826-1-IG-100UL
100 μL
360.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TP53, also known as P53 and NY-CO-13, belongs to the p53 family and has 9 isoforms. In SDS-Page, the observed molecular weight is about 53 kDa. TP53 acts as a tumor suppressor in many tumor types, including growth arrest or apoptosis depending on the physiological circumstances and cell types. It is involved in cell cycle regulation as a trans-activator that acts to negatively regulate cell division by controlling a set of genes required for this process. TP53 Localizes in the nucleus in most cells but found in the cytoplasm in some cells. (PMID: 26166714; PMID: 25225161)

Specifications

Specifications

Phospho-P53 (Ser15)
1000 μg/mL
Monoclonal
Liquid
Human
Primary
1H6G1
Western Blot, ELISA
Unconjugated
Mouse
7157
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.