missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PDPK1 (Ser241) Recombinant Rabbit Monoclonal Antibody (PDK1S241-F7), FITC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
202.00€ - 506.00€
Specifications
| Antigen | Phospho-PDPK1 (Ser241) |
|---|---|
| Clone | PDK1S241-F7 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17156928
|
Invitrogen™
MA537339 |
10 Tests |
202.00€
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17166678
![]() |
Invitrogen™
MA537090 |
100 Tests |
506.00€
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDPK1 (3 Phosphoinositide Dependent Protein Kinase 1) phosphorylates AGC kinases. PDPK1 activates conventional PKC and PKC zeta through phosphorylation of critical threonine residues in the activation loop. PDPK1 also phosphorylates Protein Kinase B (PKB) at threonine 308 in the presence of phosphatidylinositol-3,4,5-trisphosphate. Active Akt inactivates Glycogen Synthase Kinase 3 (GSK3), eventually leading to the dephosphorylation and activation of glycogen synthase, and the stimulation of glycogen synthesis. Because of the role that PDPK1 plays in insulin-induced glycogen synthesis and PKC activation, it is a potentially important target for metabolic drug research.Specifications
| Phospho-PDPK1 (Ser241) | |
| Flow Cytometry | |
| FITC | |
| Rabbit | |
| Human | |
| O15530 | |
| 5170 | |
| A synthetic phospho-peptide corresponding to residues surrounding Ser241 of human phospho PDPK1. | |
| Primary | |
| 4°C, store in dark | |
| Pdpk1 |
| PDK1S241-F7 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| 3-phosphoinositide dependent protein kinase 1; 3-phosphoinositide dependent protein kinase-1; 3-phosphoinositide-dependent protein kinase 1; 3-phosphoinositide-dependent protein kinase 2 pseudogene; HGNC:8809; hPDK1; isoenzyme 1; kinase isoenzyme 1; lipoamide; mPDK1; OTTHUMP00000174525; OTTHUMP00000205076; PDK1; PDPK1; PDPK2; PDPK2P; Pkb kinase; PkB kinase like gene 1; PkB-like 1; PRO0461; Protein kinase B kinase | |
| Pdpk1 | |
| IgG κ | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title