missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-RPA2 (Ser4, Ser8) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121338
This item is not returnable.
View return policy
Description
The target is usually found in the following locations: Nucleus, PML body. Immunogen sequence: SGFES.
Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.
Specifications
| Phospho-RPA2 (Ser4, Ser8) | |
| Polyclonal | |
| Unconjugated | |
| Rpa2 | |
| 30-kDa protein; AA409079; AI325195; AU020965; HSSB; hypothetical protein; I79_004200; p32; p32-subunit of replication protein A; p34; REPA2; Replication factor A protein 2; replication protein A 32 kDa subunit; Replication protein A 32kDa subunit; Replication protein A 34 kDa subunit; Replication protein A 34kDa subunit; replication protein A2; replication protein A2, 32kDa; replication protein A4; RFA; RF-A protein 2; Rf-A2; RP21C; RP4-547C9.3; RP-A p32; RP-A p34; RPA2; RPA32; RPA34; RPA4; RpLP1; RpP2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 59102, 6118 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P15927, Q63528 | |
| Rpa2 | |
| Synthetic Peptide. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction