missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-RRN3 (Ser649) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539814
Additional Details : Weight : 0.00900kg
Description
Required for efficient transcription initiation by RNA polymerase I. Required for the formation of the competent preinitiation complex (PIC). Dissociates from pol I as a consequence of transcription. In vitro, cannot activate transcription in a subsequent transcription reaction.Specifications
Phospho-RRN3 (Ser649) | |
Polyclonal | |
Unconjugated | |
RRN3 | |
A-270G1.2, TIFIA, RNA polymerase I-specific transcription initiation factor RRN3, TIF-IA, transcription initiation factor IA, transcription initiation factor TIF-IA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54700 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NYV6 | |
RRN3 | |
A synthetic phosphopeptide derived from human TIF-IA around the phosphorylation site of Ser649 (Q-P-SP-P-L) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |