Learn More
Phospho-Tyrosine Antibody (P-TYR-01), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB500-335MFV500
Product Code. 18750293
  • 553.00€ / 0.10mL
Description
Phospho-Tyrosine Monoclonal antibody specifically detects Phospho-Tyrosine in Non-species specific samples. It is validated for Flow Cytometry, Flow (Intracellular)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-Tyrosine
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Store at 4C in the dark.
IgG1
Flow Cytometry
P-TYR-01
Flow Cytometry, Flow (Intracellular)
2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, phosphotyrosine, pTyrosine, pY20, Tyrosine
Phosphotyrosine conjugated to BSA.
0.1 mL
Neuroscience, Phospho Specific, Signal Transduction, Tyrosine Kinases
Non-species specific