missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholipase B1 Antibody (OTI1D3), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73386APC
This item is not returnable.
View return policy
Description
Phospholipase B1 Monoclonal antibody specifically detects Phospholipase B1 in Human samples. It is validated for Western Blot
Specifications
| Phospholipase B1 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| OTI1D3 | |
| Western Blot | |
| FLJ30866, hPLB, phospholipase B1, PLBmembrane-associated | |
| Full length human recombinant protein of human PLB1(NP_694566) produced in HEK293T cell. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 151056 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction