missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholipase C like 1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06275MFV450
This item is not returnable.
View return policy
Description
Phospholipase C like 1 Polyclonal antibody specifically detects Phospholipase C like 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Phospholipase C like 1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| but catalytically inactive protein, inactive phospholipase C-like protein 1, MGC126580, MGC138190, phospholipase C, epsilon, phospholipase C-deleted in lung carcinoma, phospholipase C-like 1, phospholipase C-related but catalytically inactive protein, PLCE, PLCL, PLC-L1, PRIP | |
| Produced in rabbits immunized with E. coli-derived Human Phospholipase C like 1 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 5334 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction