missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid Tris(2-methylphenyl) Ester, TRC

Product Code. 30012680
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012680 100 g 100g
30003500 25 g 25g
18083720 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012680 Supplier Toronto Research Chemicals Supplier No. TRCP359465100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-30-8
Molecular Formula C21 H21 O4 P
Molecular Weight (g/mol) 368.36
Synonym Phosphoric acid, tris(2-methylphenyl) ester (9CI, ACI), Phosphoric acid, tri-o-tolyl ester (8CI), NSC 438, o-Cresyl phosphate, P(o-Tolyl)3, Phosflex 179C, TOCP, TOKF, TOTP, Tri-o-cresyl phosphate, Tri-o-tolyl phosphate, Tris(2-methylphenyl) phosphate, Tris(2-tolyl) phosphate, Tris(o-cresyl) phosphate, Tris(o-methylphenyl) phosphate, Tris(o-tolyl) phosphate, Tri-2-cresyl phosphate
IUPAC Name tris(2-methylphenyl) phosphate
SMILES Cc1ccccc1OP(=O)(Oc2ccccc2C)Oc3ccccc3C

Specifications

Quantity 100 g
InChI Formula InChI=1S/C21H21O4P/c1-16-10-4-7-13-19(16)23-26(22,24-20-14-8-5-11-17(20)2)25-21-15-9-6-12-18(21)3/h4-15H,1-3H3
Formula Weight 368.1177
Chemical Name or Material Tri-o-cresyl phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.