Learn More
PhosphoSHIP Rabbit, anti-Human, Unconjugated, Polyclonal, STEMCELL Technologies
Rabbit Polyclonal Antibody
Brand: Stemcell Technologies 60142
Description
- Phospho-SHIP is a highly phosphorylated form of SHIP
- SH2-containing inositol phosphatase (SHIP) is a 145 kDa hematopoietic-restricted protein that becomes tyrosine-phosphorylated and associated with the adaptor protein, Shc, following cytokine, growth factor, chemokine, and immunoreceptor stimulation
- SHIP also hydrolyzes the critical phosphatidylinositol (PI)-3-kinase (PI3K)-generated second messenger, PI-3,4,5-P3 (PIP3), to PI-3,4-P2 1,2 and therefore acts as an important negative regulator of the PI3K pathway
- This antibody reacts with mouse and human tyrosine-phosphorylated SHIP
Specifications
| PhosphoSHIP | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Granulocytes and Subsets, Lymphocytes, Myeloid Cells |
| Immunoblot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| 100 μL | |
| Human, Mouse |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.