Learn More
PhosphoSHIP Rabbit, anti-Human, Unconjugated, Polyclonal, STEMCELL Technologies
Rabbit Polyclonal Antibody
Brand:  Stemcell Technologies 60142
Encompass_Preferred
Product Code. 17129181
  • 745.20€ / 100µL
Description
  • Phospho-SHIP is a highly phosphorylated form of SHIP
  • SH2-containing inositol phosphatase (SHIP) is a 145 kDa hematopoietic-restricted protein that becomes tyrosine-phosphorylated and associated with the adaptor protein, Shc, following cytokine, growth factor, chemokine, and immunoreceptor stimulation
  • SHIP also hydrolyzes the critical phosphatidylinositol (PI)-3-kinase (PI3K)-generated second messenger, PI-3,4,5-P3 (PIP3), to PI-3,4-P2 1,2 and therefore acts as an important negative regulator of the PI3K pathway
  • This antibody reacts with mouse and human tyrosine-phosphorylated SHIP
Specifications
Specifications
PhosphoSHIP
Polyclonal
Rabbit
Primary
Granulocytes and Subsets, Lymphocytes, Myeloid Cells
Immunoblot, Flow Cytometry, Immunofluorescence, Immunoprecipitation