missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SIGLEC12 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5110369

Product Code. 17178293

  • 502.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EQKDYLLTMQ KSVTVQEGLC VSVLCSFSYP QNGWTASDPV HGYWFRAGDH VSRNIPVATN NPARAVQEET RDRFHLLGDP QNKDCTLSIR DTRESDAGTY VFCVERGNMK WNYKYDQLSV NVTASQDLLS RYRLEVPESV TVQEGLCVSV PCSVLYPHYN WTASSPVYGS WFKEGADIPW DIPVATNTPS G.

Sialic acid-binding immunoglobulin-like lectins are a family of cell surface proteins belonging to the immunoglobulin superfamily. They mediate protein-carbohydrate interactions by selectively binding to different sialic acid moieties present on glycolipids and glycoproteins. This gene encodes a member of the SIGLEC3-like subfamily of SIGLECs. Members of this subfamily are characterized by an extracellular V-set immunoglobulin-like domain followed by two C2-set immunoglobulin-like domains, and the cytoplasmic tyrosine-based motifs ITIM and SLAM-like. The encoded protein, upon tyrosine phosphorylation, has been shown to recruit the Src homology 2 domain-containing protein-tyrosine phosphatases SHP1 and SHP2. It has been suggested that the protein is involved in the negative regulation of macrophage signaling by functioning as an inhibitory receptor. This gene is located in a cluster with other SIGLEC3-like genes on 19q13.4. Alternatively spliced transcript variants encoding distinct isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SIGLEC12
Polyclonal
Unconjugated
Siglece
Cd170; MIS; mSiglec-E; Myeloid inhibitory siglec; S2V; sialic acid binding Ig like lectin 12 (gene/pseudogene); sialic acid binding Ig-like lectin 12; sialic acid binding Ig-like lectin 5; sialic acid binding Ig-like lectin E; sialic acid-binding Ig-like lectin 12; Sialic acid-binding Ig-like lectin 5; sialic acid-binding Ig-like lectin E; Sialic acid-binding Ig-like lectin-like 1; sialic acid-binding immunoglobulin-like lectin E; Siglec; SIGLEC12; siglec-12; Siglec5; Siglec-5; Siglec9; Siglece; siglec-E; Siglecl1; Siglec-L1; SIGLEC-like 1; Siglec-XII; SLG; UNQ9215/PRO34042
Rabbit
Affinity Chromatography
RUO
83382
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.98 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q91Y57
Siglece
Recombinant protein (or fragment.
100 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.