missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic-d4 Anhydride, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Phthalic Acid Anhydride-d4 |
| CAS | 75935-32-9 |
| Molecular Formula | C8 D4 O3 |
| Quantity | 1 g |
| Synonym | 1,3-Isobenzofurandione-4,5,6,7-d4 (9 CI, ACI), Phthalic-d4 anhydride (7 CI), Phthalic Anhydride-D4, Phthalic Acid Anhydride-d4 |
| InChI Formula | InChI=1 S/C8H4O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h1-4 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2C(=O)OC(=O)c2c1[2 H] |
| IUPAC Name | 4,5,6,7-tetradeuterio-2-benzofuran-1,3-dione |
| Molecular Weight (g/mol) | 152.14 |
| Formula Weight | 152.041 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction