missing translation for 'onlineSavingsMsg'
Learn More

Phthalonitrile, 98%

Product Code. 11424030
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424030 100 g 100g
11417367 250 g 250g
11407367 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424030 Supplier Thermo Scientific Alfa Aesar Supplier No. A15071.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 91-15-6 | C8H4N2 | 128.13 g/mol

Phthalonitrile is used as an intermediate for chemical synthesis in the production of building blocks for colorants and coatings, life science and agricultural chemicals. And also used as a precursor to phthalocyanine and other pigments, fluorescent brighteners, and photographic sensitizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalonitrile is used as an intermediate for chemical synthesis in the production of building blocks for colorants and coatings, life science and agricultural chemicals. And also used as a precursor to phthalocyanine and other pigments, fluorescent brighteners, and photographic sensitizers.

Solubility
Soluble in alcohol and ether.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-15-6
Molecular Formula C8H4N2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00001771
InChI Key XQZYPMVTSDWCCE-UHFFFAOYSA-N
Synonym phthalonitrile, 1,2-dicyanobenzene, o-phthalodinitrile, phthalodinitrile, 1,2-benzenedicarbonitrile, o-dicyanobenzene, 1,2-benzodinitrile, o-pdn, o-cyanobenzonitrile, o-benzenedicarbonitrile
PubChem CID 7042
IUPAC Name benzene-1,2-dicarbonitrile
SMILES N#CC1=CC=CC=C1C#N

Specifications

Melting Point 139°C to 141°C
Flash Point 162°C (323°F)
Quantity 100 g
UN Number UN3439
Beilstein 775028
Solubility Information Soluble in alcohol and ether.
Formula Weight 128.13
Percent Purity 98%
Chemical Name or Material Phthalonitrile
compliance-icons
Product Identifier
  • Phthalonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: o-Phthalodinitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.