missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI16 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12267-1-AP-150UL
This item is not returnable.
View return policy
Description
Putative serine protease inhibitor.Specifications
| PI16 | |
| Polyclonal | |
| Unconjugated | |
| PI16 | |
| CRISP 9, CRISP9, MSMBBP, peptidase inhibitor 16, PI 16, PI16, PSP94 binding protein, PSPBP, UNQ289/PRO328 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 221476 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6UXB8 | |
| PI16 | |
| PI16 Fusion Protein Ag2925 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction