missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI4KA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12411-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a phosphatidylinositol (PI) 4-kinase which catalyzes the first committed step in the biosynthesis of phosphatidylinositol 4,5-bisphosphate. The mammalian PI 4-kinases have been classified into two types, II and III, based on their molecular mass, and modulation by detergent and adenosine. The protein encoded by this gene is a type III enzyme that is not inhibited by adenosine. Two transcript variants encoding different isoforms have been described for this gene.Specifications
| PI4KA | |
| Polyclonal | |
| Unconjugated | |
| PI4KA | |
| PI4 kinase alpha, PI4K ALPHA, pi4K230, PI4KA, PIK4, PIK4CA, PtdIns 4 kinase alpha | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 224020, 5297, 64161 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| E9Q3L2, O08662, P42356 | |
| PI4KA | |
| PI4KA Fusion Protein Ag3062 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction