Description
Thermo Scientific™ MALDI Matrices are highly purified, recrystallized reagents packaged in convenient single-use tubes sufficient for analyzing up to 96 samples by matrix assisted laser desorption ionization.
The MALDI method for MS peptide analysis is now easier to perform with single-use, highly pure MALDI matrices that are exceptionally convenient to use. Matrix assisted laser desorption ionization (MALDI) matrices produce the cleanest mass spectrometric (MS) spectra when they are recrystallized and prepared daily, which is inconvenient and typically results in significant waste. The packaging of purified, recrystallized alpha-cyano-4-hydroxy-cinnamic acid (CHCA), sinapinic acid (SA) and 2,5-dihydroxybenzoic acid (DHB) MALDI matrices makes preparation of high-quality MALDI reagents easy.
Highlights:
- Highly purified, recrystallized reagent
- Packaged in convenient single-use tubes sufficient for analyzing up to 96 samples to save time and reduce waste
- Provides accurate quantity of matrix every time
Specifications
Specifications
| Description | Pierce SA MALDI Matrices |
| For Use With (Equipment) | Mass Spectrometer |
| Product Type | MALDI Matrix |
| Content And Storage | Upon receipt store product at 4°C. |
| Starting Material | Small Molecules |
| Final Product Type | Small Molecules |
| Detection Method | Mass Spectrometry |
| Workflow Step | Calibration |
| Quantity | 24 x 1 mg |
| Form | Solid |