Learn More
Thermo Scientific™ Tris
Product Code. 10443415 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10443415 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10443415 Supplier Thermo Scientific™ Supplier No. 17926
Perform a variety of protein and molecular biology methods. Thermo Scientific™ Pierce™ Tris is a standard Tris buffering reagent available in a 1kg pack.
Thermo Scientific™ Pierce Tris is a standard reagent for preparation of many molecular biology buffers.
Additional Information and Online Ordering
Thermo Scientific™ Pierce™ Tris is a standard reagent for preparation of many molecular biology buffers.
  • ≥99.8% pure
  • White crystalline powder
  • Optimal buffering between pH 7 and 9
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Product Type Tris
Purity ≥99.8%
Quantity 1 kg
Form Powder