missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIG3 Antibody (1C9), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22591AF532
This item is not returnable.
View return policy
Description
PIG3 Monoclonal specifically detects PIG3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| PIG3 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TP53I3 | |
| Recombinant human TP53I3 (1-332aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1C9 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| p53-induced gene 3 protein, PIG3quinone oxidoreductase homolog, quinone oxidoreductase PIG3, tumor protein p53 inducible protein 3, Tumor protein p53-inducible protein 3 | |
| Mouse | |
| Protein G purified | |
| Apoptosis | |
| 9540.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction