missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3CA Antibody (CL8345) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05502AF594
This item is not returnable.
View return policy
Description
PIK3CA Monoclonal antibody specifically detects PIK3CA in Human samples. It is validated for Western Blot
Specifications
| PIK3CA | |
| Monoclonal | |
| Alexa Fluor 594 | |
| EC 2.7.1, EC 2.7.1.153, MGC142161, MGC142163, p110-alpha, Phosphatidylinositol 3-Kinase catalytic subunit alpha, phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha, phosphatidylinositol 3-kinase, catalytic, alpha polypeptide, Phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha isoform, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit, alpha isoform, phosphoinositide-3-kinase, catalytic, alpha polypeptide, PI3K, PI3K-alpha, PI3-kinase p110 subunit alpha, PI3-kinase subunit alpha, PtdIns-3-kinase p110, PtdIns-3-kinase subunit alpha, PtdIns-3-kinase subunit p110-alpha | |
| This antibody was developed using a synthetic peptide derived from P42336, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Diabetes Research, mTOR Pathway, Oncogenes | |
| 5290 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL8345 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction