missing translation for 'onlineSavingsMsg'
Learn More

Pimaricin, 2.5% aqueous suspension, MP Biomedicals™

Product Code. 11403642 Shop All MP Biomedicals Products
Click to view available options
Quantity:
20 mL
Unit Size:
20mL
This item is not returnable. View return policy

Product Code. 11403642

Brand: MP Biomedicals 0215625420

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Type Pimaricin
Physical Form Suspension
CAS 7681-93-8
Color Cream
Quantity 20 mL
Molecular Formula C33H47NO13
Synonym Antibiotic A-5283, Pimafucin
InChI Key NCXMLFZGDNKEPB-UWSQUGIJSA-N
SMILES CC1CC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(CC3C(O3)C=CC(=O)O1)O)O)O)C(=O)O)OC4C(C(C(C(O4)C)O)N)O
Molecular Weight (g/mol) 665.733
PubChem CID 129317863
Formula Weight 665.75
Chemical Name or Material Pimaricin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.