missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pin1 Antibody (3G8), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-22956V
This item is not returnable.
View return policy
Description
Pin1 Monoclonal antibody specifically detects Pin1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| Pin1 | |
| Monoclonal | |
| DyLight 405 | |
| NP_006212 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 3G8 | |
| 50mM Sodium Borate | |
| dod, EC 5.2.1.8, peptidyl-prolyl cis/trans isomerase, NIMA-interacting, peptidylprolyl cis/trans isomerase, NIMA-interacting 1, peptidyl-prolyl cis-trans isomerase NIMA-interacting 1, Peptidyl-prolyl cis-trans isomerase Pin1, PPIase Pin1, prolyl isomerase, protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1, Rotamase Pin1, UBL5 | |
| Recombinant human Pin1 (1-163 aa) purified from E. coli | |
| 0.1 mL | |
| Cell Cycle and Replication, Mitotic Regulators | |
| 5300 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction