missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pin1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-53805
This item is not returnable.
View return policy
Description
Pin1 Polyclonal specifically detects Pin1 in Rat samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Pin1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| dod, EC 5.2.1.8, peptidyl-prolyl cis/trans isomerase, NIMA-interacting, peptidylprolyl cis/trans isomerase, NIMA-interacting 1, peptidyl-prolyl cis-trans isomerase NIMA-interacting 1, Peptidyl-prolyl cis-trans isomerase Pin1, PPIase Pin1, prolyl isomerase, protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1, Rotamase Pin1, UBL5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Expected from sequence similarity: Human, Mouse and Dog. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.2 - 0.6 ug/ml, Peptide ELISA Detection limit 1:128000 | |
| NP_006212.1 | |
| PIN1 | |
| Peptide with sequence C-KHSQSRRPSSWRQEKITR corresponding to internal region according to NP_006212.1. | |
| 0.1 mg | |
| Cell Cycle and Replication, Mitotic Regulators | |
| 5300 | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction