Learn More
PIN4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15789-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the parvulin subfamily of the peptidyl-prolyl cis/trans isomerase protein family. The encoded protein catalyzes the isomerization of peptidylprolyl bonds, and may play a role in the cell cycle, chromatin remodeling.Specifications
| PIN4 | |
| Polyclonal | |
| Unconjugated | |
| PIN4 | |
| EPVH, hEPVH, hPar14, hPar17, PAR14, PAR17, Parvulin 14, Parvulin 17, PIN4, PPIase Pin4, Rotamase Pin4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5303, 684441, 69713 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9CWW6, Q9Y237 | |
| PIN4 | |
| PIN4 Fusion Protein Ag8489 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |