missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Pindolol-d7 (iso-propyl-d7), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Pindolol-d7 |
| CAS | 1185031-19-9 |
| Molecular Formula | C14 D7 H13 N2 O2 |
| Quantity | 0.005 g |
| Synonym | 1-(1,1,1,2,3,3,3-Heptadeuteriopropan-2-ylamino)-3-(1 H-indol-4-yloxy)propan-2-ol, (±)-Pindolol-d7 (iso-propyl-d7), Pindolol-d7, Pindolol-D7 (N-isopropyl-D7), 1-(1 H-Indol-4-yloxy)-3-[[1-(methyl-d3)ethyl-1,2,2,2-d4]amino]-2-propanol |
| InChI Formula | InChI=1 S/C14H20N2O2/c1-10(2)16-8-11(17)9-18-14-5-3-4-13-12(14)6-7-15-13/h3-7,10-11,15-17 H,8-9H2,1-2H3/i1D3,2D3,10 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(NCC(O)COc1cccc2[nH]ccc12)C([2 H])([2 H])[2 H] |
| IUPAC Name | 1-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)-3-(1 H-indol-4-yloxy)propan-2-ol |
| Molecular Weight (g/mol) | 255.364 |
| Formula Weight | 255.196 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction