missing translation for 'onlineSavingsMsg'
Learn More

Pinobanksin, TRC

Product Code. 17904995
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17904995 25 mg 25mg
17952565 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17904995 Supplier Toronto Research Chemicals Supplier No. TRCP46835025MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 548-82-3
Molecular Formula C15H12O5
Molecular Weight (g/mol) 272.25
Synonym (2R,3R)-2,3-Dihydro-3,5,7-trihydroxy-2-phenyl-4H-1-benzopyran-4-one, (2R-trans)-2,3-Dihydro-3,5,7-trihydroxy-2-phenyl-4H-1-benzopyran-4-one, 3,5,7-Trihydroxyflavanone, (2R,3R)-Pinobanksin, 3,5,7-Trihydroxyflavanone, Dihydrogalangin
IUPAC Name (2R,3R)-3,5,7-trihydroxy-2-phenyl-2,3-dihydrochromen-4-one
SMILES O[C@@H]1[C@H](Oc2cc(O)cc(O)c2C1=O)c3ccccc3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H12O5/c16-9-6-10(17)12-11(7-9)20-15(14(19)13(12)18)8-4-2-1-3-5-8/h1-7,14-17,19H/t14-,15+/m0/s1
Formula Weight 272.0685
Chemical Name or Material Pinobanksin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.