missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIP Antibody (PIP/9076R), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24017AF647
This item is not returnable.
View return policy
Description
PIP Monoclonal antibody specifically detects PIP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PIP | |
| Monoclonal | |
| Alexa Fluor 647 | |
| GCDFP15prolactin-inducible protein, GCDFP-15SABP, GPIP4gp17, Gross cystic disease fluid protein 15, prolactin-induced proteinSecretory actin-binding protein | |
| Recombinant fragment (around aa 1-146) of human PIP protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Cellular Markers, Signal Transduction | |
| 5304 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| PIP/9076R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction