Learn More
Brady™ Pipe Markers - with Holder Water
Product Code. 12539747 Shop All Brady Products
Change view
Click to view available options
Display Text:
Eau de chauffe
Eau glacée
Eau pluviale
Unit Size:
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Unit Size
12539747 Eau de chauffe Pack of 4
12519747 Eau glacée Pack of 4
12549747 Eau pluviale Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12539747 Supplier Brady™ Supplier No. N003379
  • ONLINE EXCLUSIVE PRICE
    15.65€
    14.14€ / Pack of 4
    Save 1.51€
    9% Off
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC
Description Pipe Markers - Eau de chauffe
Adhesive Type Permanent Acrylic
Coating Polyester
Color White on Green
Display Text
Finish Gloss
Format Card
Material Polyester
Product Type Pipe Marker
Sterility Non-sterile
Display Text Eau de chauffe
For Use With (Application) Pipe and valve marking
Labels per Sheet 4
Languages French
No. per Pack 4 Markers/Card
Quantity 4 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (L x W) 26 x 250 mm
Length (English) 1.02 in.
Length (Metric) 26 mm
Width (English) 9.84 in.
Width (Metric) 250 mm
Alternate Part Number E.DECHAUFFE250X26CARD-T1-P16