missing translation for 'onlineSavingsMsg'
Learn More

Piperacetazine, TRC

Product Code. 30422730
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422730 100 mg -
30420538 2.5 g 2.5g
30424040 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30422730 Supplier Toronto Research Chemicals Supplier No. TRCP479975100MG
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3819-00-9
Molecular Formula C24 H30 N2 O2 S
Molecular Weight (g/mol) 410.57
Synonym Ethanone, 1-[10-[3-[4-(2-hydroxyethyl)-1-piperidinyl]propyl]-10H-phenothiazin-2-yl]-, Ketone, 10-[3-[4-(2-hydroxyethyl)piperidino]propyl]phenothiazin-2-yl methyl (6CI,7CI,8CI), 10-[3-[4-(2-Hydroxyethyl)piperidino]propyl]phenothiazin-2-yl methyl ketone, PC 1421, Piperacetazine, Psymod, Quide, SC 9794, 2-Acetyl-10-[3-[4-(beta-hydroxyethyl)piperidino]propyl]phenothiazine
IUPAC Name 1-[10-[3-[4-(2-hydroxyethyl)piperidin-1-yl]propyl]phenothiazin-2-yl]ethanone
SMILES CC(=O)c1ccc2Sc3ccccc3N(CCCN4CCC(CCO)CC4)c2c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C24H30N2O2S/c1-18(28)20-7-8-24-22(17-20)26(21-5-2-3-6-23(21)29-24)13-4-12-25-14-9-19(10-15-25)11-16-27/h2-3,5-8,17,19,27H,4,9-16H2,1H3
Formula Weight 410.2028
Chemical Name or Material Piperacetazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.