Learn More
Piperlongumine, TRC
Specifications
Specifications
| Chemical Name or Material | Piperlongumine |
| CAS | 20069-09-4 |
| Molecular Formula | C17 H19 N O5 |
| Quantity | 25 mg |
| Synonym | 5,6-Dihydro-1-[(2E)-1-oxo-3-(3,4,5-trimethoxyphenyl)-2-propen-1-yl]-2(1H)-pyridinone |
| InChI Formula | InChI=1S/C17H19NO5/c1-21-13-10-12(11-14(22-2)17(13)23-3)7-8-16(20)18-9-5-4-6-15(18)19/h4,6-8,10-11H,5,9H2,1-3H3/b8-7+ |
| SMILES | COc1cc(\C=C\C(=O)N2CCC=CC2=O)cc(OC)c1OC |
| IUPAC Name | 1-[(E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoyl]-2,3-dihydropyridin-6-one |
| Molecular Weight (g/mol) | 317.34 |
| Formula Weight | 317.1263 |