Learn More
Gilson™ PIPETMAN™ G
GREENER_CHOICE
Product Code. 16313186 Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
1 to 10 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
Number of Channels:
8
12
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Number of Channels unitSize
16313186 1 to 10 μL 8 Each
16323186 1 to 10 μL 12 Each
16333186 2 to 20 μL 8 Each
16343186 2 to 20 μL 12 Each
16353186 20 to 200 μL 8 Each
16363186 20 to 200 μL 12 Each
16373186 30 to 300 μL 8 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 16313186 Supplier Gilson™ Supplier No. F144068
  • ONLINE EXCLUSIVE PRICE
    864.46€
    841.05€ / Pack of 1
    Save 23.41€
    2% Off
View all promotions
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
PIPETMAN™ G offers a low pipetting force and hours of comfort due to a redesigned piston assembly system.
  • With a redesigned piston assembly system and high quality piston grease, PIPETMAN™ G offers hours of comfortable pipetting
  • PIPETMAN™ G relies on the legendary accuracy, precision, and reliability of PIPETMAN™ with improved pipetting comfort
  • Made of PVDF and stainless steel
  • Offers premium durability
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Declaration of Conformity
Autoclavable Not Autoclavable
Color Red
Material Metal Ejector
Model P8x10G
Number of Channels 8
Product Line Pipetman G
Type Manual Air Displacement Pipette
Volume (Metric) 1 to 10 μL
Warranty 3 Years
Length (English) 5.98 in.
Width (English) 12.87 in.
Height (English) 1.54 in.