missing translation for 'onlineSavingsMsg'
Learn More

Piretanide, TRC

Product Code. 17904165
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17904165 1 mg 1mg
17910675 2.5 mg 2.5mg
17914485 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17904165 Supplier Toronto Research Chemicals Supplier No. TRCP5087001MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55837-27-9
Molecular Formula C17 H18 N2 O5 S
Molecular Weight (g/mol) 362.4
Synonym Piretanide, 4-Phenoxy-3-(pyrrolidin-1-yl)-5-sulfamoylbenzoic acid
IUPAC Name 4-phenoxy-3-pyrrolidin-1-yl-5-sulfamoylbenzoic acid
SMILES NS(=O)(=O)c1cc(cc(N2CCCC2)c1Oc3ccccc3)C(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C17H18N2O5S/c18-25(22,23)15-11-12(17(20)21)10-14(19-8-4-5-9-19)16(15)24-13-6-2-1-3-7-13/h1-3,6-7,10-11H,4-5,8-9H2,(H,20,21)(H2,18,22,23)
Formula Weight 362.0936
Percent Purity >95%
Chemical Name or Material Piretanide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.