Learn More
Piretanide, TRC
Product Code. 17914485
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17914485 5 mg 5mg
17904165 1 mg 1mg
17910675 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17914485 Supplier Toronto Research Chemicals Supplier No. TRCP5087005MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 55837-27-9
Molecular Formula C17 H18 N2 O5 S
Molecular Weight (g/mol) 362.4
Synonym Piretanide, 4-Phenoxy-3-(pyrrolidin-1-yl)-5-sulfamoylbenzoic acid
IUPAC Name 4-phenoxy-3-pyrrolidin-1-yl-5-sulfamoylbenzoic acid
SMILES NS(=O)(=O)c1cc(cc(N2CCCC2)c1Oc3ccccc3)C(=O)O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C17H18N2O5S/c18-25(22,23)15-11-12(17(20)21)10-14(19-8-4-5-9-19)16(15)24-13-6-2-1-3-7-13/h1-3,6-7,10-11H,4-5,8-9H2,(H,20,21)(H2,18,22,23)
Formula Weight 362.0936
Percent Purity >95%
Chemical Name or Material Piretanide
Grade HPLC