missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piroctone Olamine, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Piroctone Olamine |
| CAS | 68890-66-4 |
| Molecular Formula | C14 H23 N O2 . C2 H7 N O |
| Quantity | 250 mg |
| Synonym | Ethanol','2-amino-','compd. with 1-hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1H)-pyridinone (1:1) (9CI), 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2-pyridone monoethanolamine salt, Octopirox, Octopyrox, Piroctone ethanolamine salt, Piroctone olamine |
| InChI Formula | InChI=1S/C14H23NO2.C2H7NO/c1-10-6-12(15(17)13(16)8-10)7-11(2)9-14(3,4)5;3-1-2-4/h6,8,11,17H,7,9H2,1-5H3;4H,1-3H2 |
| SMILES | CC(CC1=CC(=CC(=O)N1O)C)CC(C)(C)C.NCCO |
| IUPAC Name | 2-aminoethanol;1-hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)pyridin-2-one |
| Molecular Weight (g/mol) | 298.42 |
| Formula Weight | 298.2256 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction