missing translation for 'onlineSavingsMsg'
Learn More

Piroctone Olamine, TRC

Product Code. 30426368
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30426368

Brand: Toronto Research Chemicals TRCP50940050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Piroctone Olamine
CAS 68890-66-4
Molecular Formula C14 H23 N O2 . C2 H7 N O
Quantity 50 mg
Synonym Ethanol','2-amino-','compd. with 1-hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1H)-pyridinone (1:1) (9CI), 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2-pyridone monoethanolamine salt, Octopirox, Octopyrox, Piroctone ethanolamine salt, Piroctone olamine
InChI Formula InChI=1S/C14H23NO2.C2H7NO/c1-10-6-12(15(17)13(16)8-10)7-11(2)9-14(3,4)5;3-1-2-4/h6,8,11,17H,7,9H2,1-5H3;4H,1-3H2
SMILES CC(CC1=CC(=CC(=O)N1O)C)CC(C)(C)C.NCCO
IUPAC Name 2-aminoethanol;1-hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)pyridin-2-one
Molecular Weight (g/mol) 298.42
Formula Weight 298.2256
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.