missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PITX2 Antibody (OTI1H10), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-45000-30ul
This item is not returnable.
View return policy
Description
PITX2 Monoclonal antibody specifically detects PITX2 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| PITX2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1H10 | |
| Western Blot 1:2000 | |
| all1-responsive gene 1, ALL1-responsive protein ARP1, ARP1MGC20144, Homeobox protein PITX2, IDG2, IGDS, IGDS2, IHG2, IRID2, Otlx2, paired-like homeodomain 2, Paired-like homeodomain transcription factor 2MGC111022, pituitary homeo box 2, pituitary homeobox 2, PTX2, RGSRIEG1Brx1, RIEG, RIEG bicoid-related homeobox transcription factor, rieg bicoid-related homeobox transcription factor 1, RS, solurshin | |
| Human recombinant protein fragment corresponding to amino acids 1-200 of human PITX2 (NP_700475) produced in E.coli. | |
| 30 μL | |
| Stem Cell Markers | |
| 5308 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction