missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIWIL4 Monoclonal Antibody (626001)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561649
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
PIWIL4 belongs to the Argonaute family of proteins, which function in development and maintenance of germline stem cells (Sasaki et al., 2003 [PubMed 12906857]).
Specifications
| PIWIL4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q7Z3Z4 | |
| PIWIL4 | |
| E. coli-derived recombinant human PIWIL4, Ser102-Gln237. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry | |
| 626001 | |
| Unconjugated | |
| PIWIL4 | |
| 9230101H05Rik; HIWI2; mAgo5; Miwi2; PIWI; piwi like RNA-mediated gene silencing 4; Piwil4; piwi-like 4; piwi-like homolog 4; piwi-like protein 4; piwi-like RNA-mediated gene silencing 4; testis tissue sperm-binding protein Li 85P | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 143689 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction